accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111109-77-4

H3COOOCH3

CAS RN 111109-77-4

IUPAC Name: 1-methoxy-3-(3-methoxypropoxy)propane
Synonyms: dipropylene glycol dimethyl ether
Molecular Weight (g/mol): 162.23
Molecular Formula: C8H18O3
InChi Key: KWDCKLXGUZOEGM-UHFFFAOYSA-N
SMILES: COCCCOCCCOC
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)