accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1118-89-4

IUPAC Name: hydrogen 1,5-diethyl (2S)-2-aminopentanedioate chloride
Synonyms: hydrogen ethyl glutamate chloride
Molecular Weight (g/mol): 239.7
Molecular Formula: C9H18ClNO4
InChi Key: WSEQLMQNPBNMSL-FJXQXJEOSA-N
SMILES: [H+].[Cl-].CCOC(=O)CC[C@H](N)C(=O)OCC
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (2)
Quantity 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)