Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid diethyl ester hydrochloride, 98%, Thermo Scientific™

Catalog No. AC459260050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC459260050 Supplier Thermo Scientific Chemicals Supplier No. 459260050
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1118-89-4
Molecular Formula C9H17NO4·HCl
Molecular Weight (g/mol) 239.7
InChI Key WSEQLMQNPBNMSL-FJXQXJEOSA-N
Synonym l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
PubChem CID 73960
IUPAC Name diethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CCOC(=O)CCC(C(=O)OCC)N.Cl

Specifications

Melting Point 108.0°C to 110.0°C
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information (5% in water) Clear colorless
Formula Weight 239.7
Percent Purity 98%
Chemical Name or Material L-Glutamic acid diethyl ester hydrochloride
EINECSNumber 214-270-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.