Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid diethyl ester hydrochloride, 98%

Catalog No. AAH5955114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5955114 25 g
AAH5955106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5955114 Supplier Thermo Scientific Chemicals Supplier No. H5955114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1118-89-4 | C9H18ClNO4 | 239.696 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-89-4
Molecular Formula C9H18ClNO4
Molecular Weight (g/mol) 239.696
MDL Number MFCD00012509
InChI Key WSEQLMQNPBNMSL-FJXQXJEOSA-N
Synonym l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
PubChem CID 73960
IUPAC Name diethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CCOC(=O)CCC(C(=O)OCC)N.Cl

Specifications

Melting Point 108°C to 110°C
Linear Formula C2H5OCOCH2CH2CH(NH2)COOC2H5·HCl
Beilstein 214-270-
Quantity 25 g
Formula Weight 239.7
Optical Rotation +22° (c=1 in H2O)
Percent Purity 98%
Chemical Name or Material L-Glutamic acid diethyl ester hydrochloride
EINECSNumber 214-270-4
RTECSNumber MA1252280
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.