accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-00-5

IUPAC Name: dodecyltrimethylazanium chloride
Synonyms: laurtrimonium chloride
Molecular Weight (g/mol): 263.89
Molecular Formula: C15H34ClN
InChi Key: DDXLVDQZPFLQMZ-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (7)
Quantity 
  • (5)
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (4)
  • (4)
  • (2)
  • (4)
  • (7)
  • (5)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)