Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyltrimethylammonium Chloride 98.0+%, TCI America™
SDP

Catalog No. I0453500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0453500G 500 g
I045325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0453500G Supplier TCI America Supplier No. I0453500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-00-5
Molecular Formula C15H34ClN
Molecular Weight (g/mol) 263.894
MDL Number MFCD00041974
InChI Key DDXLVDQZPFLQMZ-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
PubChem CID 8152
IUPAC Name dodecyl(trimethyl)azanium;chloride
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point 246°C
Color White
pH 4.0 to 7.5 (50g/L H2O 25°C)
Quantity 500 g
UN Number 3077
Formula Weight 263.89
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecyltrimethylammonium Chloride [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-0184&(9)-1971
RTECSNumber JR2122500
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.