Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyltrimethylammonium Chloride, 50% Soln, MP Biomedicals

Catalog No. ICN204484 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204484 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204484 Supplier MP Biomedicals, Inc Supplier No. 0520448480
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cationic surfactant suitable for research and laboratory applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-00-5
Molecular Formula C15H34ClN
Molecular Weight (g/mol) 263.894
InChI Key DDXLVDQZPFLQMZ-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
PubChem CID 8152
IUPAC Name dodecyl(trimethyl)azanium;chloride
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Cl-]

Specifications

Quantity 100 g
Formula Weight 263.9
Percent Purity 50%
Physical Form Liquid
Chemical Name or Material Dodecyltrimethylammonium Chloride, 50% Soln
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.