CAS RN 112-39-0
Methyl palmitate, 97%
CAS: 112-39-0 Molecular Formula: C17H34O2 Molecular Weight (g/mol): 270.457 MDL Number: MFCD00008994 InChI Key: FLIACVVOZYBSBS-UHFFFAOYSA-N Synonym: methyl palmitate,palmitic acid methyl ester,hexadecanoic acid, methyl ester,palmitic acid, methyl ester,uniphat a60,metholene 2216,methyl n-hexadecanoate,hexadecanoic acid methyl ester,n-hexadecanoic acid methyl ester,unii-dpy8vcm98i PubChem CID: 8181 ChEBI: CHEBI:69187 IUPAC Name: methyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OC
Methyl palmitate, 99%, analytical standard for GC
CAS: 112-39-0 Molecular Formula: C17H34O2 Molecular Weight (g/mol): 270.45 InChI Key: FLIACVVOZYBSBS-UHFFFAOYSA-N Synonym: methyl palmitate,palmitic acid methyl ester,hexadecanoic acid, methyl ester,palmitic acid, methyl ester,uniphat a60,metholene 2216,methyl n-hexadecanoate,hexadecanoic acid methyl ester,n-hexadecanoic acid methyl ester,unii-dpy8vcm98i PubChem CID: 8181 ChEBI: CHEBI:69187 IUPAC Name: methyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OC
Methyl palmitate, 95%
CAS: 112-39-0 Molecular Formula: C17H34O2 Molecular Weight (g/mol): 270.45 MDL Number: MFCD00008994 InChI Key: FLIACVVOZYBSBS-UHFFFAOYSA-N Synonym: methyl palmitate,palmitic acid methyl ester,hexadecanoic acid, methyl ester,palmitic acid, methyl ester,uniphat a60,metholene 2216,methyl n-hexadecanoate,hexadecanoic acid methyl ester,n-hexadecanoic acid methyl ester,unii-dpy8vcm98i PubChem CID: 8181 ChEBI: CHEBI:69187 IUPAC Name: methyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OC
TraceCERT™ Methyl Palmitate, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.
Methyl Palmitate 97.0+%, TCI America™
CAS: 112-39-0 Molecular Formula: C17H34O2 Molecular Weight (g/mol): 270.457 MDL Number: MFCD00008994 InChI Key: FLIACVVOZYBSBS-UHFFFAOYSA-N Synonym: methyl palmitate,palmitic acid methyl ester,hexadecanoic acid, methyl ester,palmitic acid, methyl ester,uniphat a60,metholene 2216,methyl n-hexadecanoate,hexadecanoic acid methyl ester,n-hexadecanoic acid methyl ester,unii-dpy8vcm98i PubChem CID: 8181 ChEBI: CHEBI:69187 IUPAC Name: methyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OC
Methyl Palmitate 99.5+%, TCI America™
CAS: 112-39-0 Molecular Formula: C17H34O2 Molecular Weight (g/mol): 270.457 MDL Number: MFCD00008994 InChI Key: FLIACVVOZYBSBS-UHFFFAOYSA-N Synonym: methyl palmitate,palmitic acid methyl ester,hexadecanoic acid, methyl ester,palmitic acid, methyl ester,uniphat a60,metholene 2216,methyl n-hexadecanoate,hexadecanoic acid methyl ester,n-hexadecanoic acid methyl ester,unii-dpy8vcm98i PubChem CID: 8181 ChEBI: CHEBI:69187 IUPAC Name: methyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OC
MP Biomedicals, Inc Palmitic Acid Methyl Ester, 99+%, MP Biomedicals™
CAS: 112-39-0 Molecular Formula: C17H34O2 Molecular Weight (g/mol): 270.457 InChI Key: FLIACVVOZYBSBS-UHFFFAOYSA-N Synonym: methyl palmitate,palmitic acid methyl ester,hexadecanoic acid, methyl ester,palmitic acid, methyl ester,uniphat a60,metholene 2216,methyl n-hexadecanoate,hexadecanoic acid methyl ester,n-hexadecanoic acid methyl ester,unii-dpy8vcm98i PubChem CID: 8181 ChEBI: CHEBI:69187 IUPAC Name: methyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OC
Methyl Palmitate, SPEX CertiPrep™
CAS: 112-39-0 Molecular Formula: C17H34O2 Molecular Weight (g/mol): 270.457 InChI Key: FLIACVVOZYBSBS-UHFFFAOYSA-N PubChem CID: 8181 ChEBI: CHEBI:69187 IUPAC Name: methyl hexadecanoate SMILES: CCCCCCCCCCCCCCCC(=O)OC