accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-59-4

IUPAC Name: 2-[2-(hexyloxy)ethoxy]ethan-1-ol
Synonyms: 2-[2-(hexyloxy)ethoxy]ethanol
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H22O3
InChi Key: GZMAAYIALGURDQ-UHFFFAOYSA-N
SMILES: CCCCCCOCCOCCO
Boiling Point 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)