accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-77-6

IUPAC Name: (9Z)-octadec-9-enoyl chloride
Synonyms: (9Z)-octadec-9-enoyl chloride
Molecular Weight (g/mol): 300.91
Molecular Formula: C18H33ClO
InChi Key: MLQBTMWHIOYKKC-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(Cl)=O
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)