Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleoyl Chloride (Mixture with ca 35-40% Elaidic Acid Chloride) (Tech.), ca 30%, Thermo Scientific™

Catalog No. AC416521000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC416521000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC416521000 Supplier Thermo Scientific Chemicals Supplier No. 416521000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-77-6
Molecular Formula C18H33ClO
Molecular Weight (g/mol) 300.91
MDL Number MFCD00134332
InChI Key MLQBTMWHIOYKKC-KTKRTIGZSA-N
Synonym elaidoylchloride, elaidoyl chloride, 9-octadecenoyl chloride, oleoylchloride, elaidic acid chloride, trans-9-octadecenoyl chloride, e-octadec-9-enoyl chloride, 9e-octadec-9-enoyl chloride
PubChem CID 5702750
IUPAC Name (9Z)-octadec-9-enoyl chloride
SMILES CCCCCCCC\C=C/CCCCCCCC(Cl)=O

Specifications

CAS Min % 25.0
CAS Max % 100.0
Color Brown
Density 0.91
Infrared Spectrum Authentic
Linear Formula CH3(CH2)7CHCH(CH2)7COCl
Quantity 100 g
Formula Weight 300.9
Percent Purity ≥25%
Grade Technical
Physical Form Liquid
Chemical Name or Material Oleoyl chloride, (mixture with ca. 35-40%elaidic acid chloride), tech., ca. 30%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.