CAS RN 112-85-6
Behenic acid, tech. 85%
CAS: 112-85-6 Molecular Formula: C22H44O2 Molecular Weight (g/mol): 340.592 MDL Number: MFCD00002807 InChI Key: UKMSUNONTOPOIO-UHFFFAOYSA-N Synonym: behenic acid,1-docosanoic acid,n-docosanoic acid,hydrofol acid 560,docosoic acid,hydrofol 2022-55,glycon b-70,behensaeure,docosanoicacid,docosansaeure PubChem CID: 8215 ChEBI: CHEBI:28941 IUPAC Name: docosanoic acid SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)O
Docosanoic acid, 85%, technical
CAS: 112-85-6 Molecular Formula: C22H44O2 Molecular Weight (g/mol): 340.58 MDL Number: MFCD00002807 InChI Key: UKMSUNONTOPOIO-UHFFFAOYSA-N Synonym: behenic acid,1-docosanoic acid,n-docosanoic acid,hydrofol acid 560,docosoic acid,hydrofol 2022-55,glycon b-70,behensaeure,docosanoicacid,docosansaeure PubChem CID: 8215 ChEBI: CHEBI:28941 IUPAC Name: docosanoic acid SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)O
Behenic Acid, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™
Behenic acid is a saturated fatty acid with cholesterol-raising ability in humans.
Behenic acid, ≥99% (GC), MilliporeSigma™ Supelco™
Behenic acid is a saturated fatty acid with cholesterol-raising ability in humans.
Behenic Acid 99.0+%, TCI America™
CAS: 112-85-6 Molecular Formula: C22H44O2 Molecular Weight (g/mol): 340.592 MDL Number: MFCD00002807 InChI Key: UKMSUNONTOPOIO-UHFFFAOYSA-N Synonym: behenic acid,1-docosanoic acid,n-docosanoic acid,hydrofol acid 560,docosoic acid,hydrofol 2022-55,glycon b-70,behensaeure,docosanoicacid,docosansaeure PubChem CID: 8215 ChEBI: CHEBI:28941 IUPAC Name: docosanoic acid SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)O
Behenic Acid 95.0+%, TCI America™
CAS: 112-85-6 Molecular Formula: C22H44O2 Molecular Weight (g/mol): 340.592 MDL Number: MFCD00002807 InChI Key: UKMSUNONTOPOIO-UHFFFAOYSA-N Synonym: behenic acid,1-docosanoic acid,n-docosanoic acid,hydrofol acid 560,docosoic acid,hydrofol 2022-55,glycon b-70,behensaeure,docosanoicacid,docosansaeure PubChem CID: 8215 ChEBI: CHEBI:28941 IUPAC Name: docosanoic acid SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)O
Behenic Acid 80.0+%, TCI America™
CAS: 112-85-6 Molecular Formula: C22H44O2 Molecular Weight (g/mol): 340.592 MDL Number: MFCD00002807 InChI Key: UKMSUNONTOPOIO-UHFFFAOYSA-N Synonym: behenic acid,1-docosanoic acid,n-docosanoic acid,hydrofol acid 560,docosoic acid,hydrofol 2022-55,glycon b-70,behensaeure,docosanoicacid,docosansaeure PubChem CID: 8215 ChEBI: CHEBI:28941 IUPAC Name: docosanoic acid SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)O
Sigma Aldrich Behenic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Behenic Acid, 85%, MP Biomedicals
CAS: 112-85-6 Molecular Formula: C22H44O2 Molecular Weight (g/mol): 340.592 InChI Key: UKMSUNONTOPOIO-UHFFFAOYSA-N Synonym: behenic acid,1-docosanoic acid,n-docosanoic acid,hydrofol acid 560,docosoic acid,hydrofol 2022-55,glycon b-70,behensaeure,docosanoicacid,docosansaeure PubChem CID: 8215 ChEBI: CHEBI:28941 IUPAC Name: docosanoic acid SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)O