Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Behenic acid, tech. 85%

Catalog No. AAA128500B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA128500B 1000 g
AAA1285030 250 g
AAA128500I 5000 g
AAA1285018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA128500B Supplier Thermo Scientific Chemicals Supplier No. A128500B
  • $164.00
    $141.65 / Each of 1

    Save $22.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 112-85-6 | C22H44O2 | 340.592 g/mol

Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

Solubility
Soluble in DMF (∼3 mg/ml), hot methanol, water (0.15 mg/ml at 25°C), chloroform, and ethanol (2.18 mg/ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, reducing agents and bases. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-85-6
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.592
MDL Number MFCD00002807
InChI Key UKMSUNONTOPOIO-UHFFFAOYSA-N
Synonym behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure
PubChem CID 8215
ChEBI CHEBI:28941
IUPAC Name docosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 74°C to 78°C
Boiling Point 306°C (60 mmHg)
Quantity 1000 g
Beilstein 1792887
Merck Index 14,1023
Solubility Information Soluble in DMF (∽3mg/ml),hot methanol,water (0.15mg/ml at 25°C),chloroform,and ethanol (2.18mg/ml at 25°C).
Formula Weight 340.6
Percent Purity 85%
Grade Technical
Chemical Name or Material Behenic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-010-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.