accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOO

CAS RN 1121-03-5

IUPAC Name: 3-methyl-1,2λ⁶-oxathiolane-2,2-dione
Synonyms: 3-methyl-1,2λ⁶-oxathiolane-2,2-dione
Molecular Weight (g/mol): 136.17
Molecular Formula: C4H8O3S
InChi Key: VWEYDBUEGDKEHC-UHFFFAOYNA-N
SMILES: CC1CCOS1(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results