Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Butanesultone 98.0+%, TCI America™
SDP

Catalog No. B38825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38825G Supplier TCI America Supplier No. B38825G

missing translation for 'validMainframeMsg'

Specifications

CAS 1121-03-5
Boiling Point 150°C
Molecular Formula C4H8O3S
MDL Number MFCD01941639
Quantity 5 g
Synonym 3-Methyl-1,2-oxathiolane 2,2-Dioxide, 1-Methyl-1,3-propanesultone
InChI Key VWEYDBUEGDKEHC-UHFFFAOYNA-N
SMILES CC1CCOS1(=O)=O
IUPAC Name 3-methyl-1,2λ⁶-oxathiolane-2,2-dione
Molecular Weight (g/mol) 136.17
PubChem CID 70716
Formula Weight 136.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Butanesultone
Show More Show Less
EINECSNumber (5)-3570
RTECSNumber RP5480000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.