CAS RN 1121-97-7
IUPAC Name:
(5-methyl-1,3-dioxan-5-yl)methanol
Synonyms:
(5-methyl-1,3-dioxan-5-yl)methanol
Molecular Weight (g/mol):
132.16
Molecular Formula:
C6H12O3
InChi Key:
XZDDZTGNCUMJHI-UHFFFAOYSA-N
SMILES:
CC1(CO)COCOC1
1
–
1
of
1
results
1
5-Hydroxymethyl-5-methyl-1,3-dioxane 97%, Thermo Scientific™
CAS: 1121-97-7 Molecular Formula: C6H12O3 Molecular Weight (g/mol): 132.16 MDL Number: MFCD00023825 InChI Key: XZDDZTGNCUMJHI-UHFFFAOYSA-N Synonym: 5-hydroxymethyl-5-methyl-1,3-dioxane,5-methyl-1,3-dioxan-5-yl methanol,5-methyl-1,3-dioxane-5-methanol,m-dioxane-5-methanol, 5-methyl,1,3-dioxane-5-methanol, 5-methyl,1,3-dioxane-5-methanol,5-methyl,5-methyl-1,3-dioxan-5-yl methan-1-ol,5-19-02-00358 beilstein handbook reference,5-methyl-m-dioxane-5-methanol PubChem CID: 14278 IUPAC Name: (5-methyl-1,3-dioxan-5-yl)methanol SMILES: CC1(CO)COCOC1