accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1122-96-9

IUPAC Name: 4-methoxypyridin-1-ium-1-olate
Synonyms: 4-methoxypyridin-1-ium-1-olate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: BOFAIBPJCWFJFT-UHFFFAOYSA-N
SMILES: COC1=CC=[N+]([O-])C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)