Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxypyridine-N-oxide, 95%, Thermo Scientific™

Catalog No. AAA1171306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1171306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1171306 Supplier Thermo Scientific Chemicals Supplier No. A1171306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-96-9
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00006207
InChI Key BOFAIBPJCWFJFT-UHFFFAOYSA-N
Synonym 4-methoxypyridine n-oxide, 4-methoxypyridine 1-oxide, 4-methoxypyridine-n-oxide, pyridine, 4-methoxy-, 1-oxide, 4-methoxypyridin-1-ium-1-olate, 4-methoxypyridine-n-oxide hydrate, pubchem2587, acmc-1buuj, 4-methoxypyridinen-oxide, 4-methoxypiridine-n-oxide
PubChem CID 70743
IUPAC Name 4-methoxy-1-oxidopyridin-1-ium
SMILES COC1=CC=[N+](C=C1)[O-]

Specifications

Melting Point 73°C to 79°C
Quantity 5 g
Formula Weight 125.13
Percent Purity 95%
Chemical Name or Material 4-Methoxypyridine-N-oxide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.