CAS RN 1125758-85-1
IUPAC Name:
N'-cyano-N''-(1-phenylethyl)-N-(quinolin-5-yl)guanidine
Synonyms:
N'-cyano-N''-(1-phenylethyl)-N-(quinolin-5-yl)guanidine
Molecular Weight (g/mol):
315.38
Molecular Formula:
C19H17N5
InChi Key:
PQYCRDPLPKGSME-UHFFFAOYNA-N
SMILES:
CC(N=C(NC#N)NC1=C2C=CC=NC2=CC=C1)C1=CC=CC=C1