accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNONNCH3H3CN(E/Z)

CAS RN 1128165-88-7

IUPAC Name: 2-cyano-3-{1-[3-(dimethylamino)propyl]-1H-indol-3-yl}-N-octylprop-2-enamide
Synonyms: 2-cyano-3-{1-[3-(dimethylamino)propyl]indol-3-yl}-N-octylprop-2-enamide
Molecular Weight (g/mol): 408.59
Molecular Formula: C25H36N4O
InChi Key: MYIMRONQBLZJFI-UHFFFAOYSA-N
SMILES: CCCCCCCCNC(=O)C(=CC1=CN(CCCN(C)C)C2=CC=CC=C12)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)