accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)OHHOHNOOO

CAS RN 112883-29-1

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxyphenyl)propanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 403.43
Molecular Formula: C24H21NO5
InChi Key: SWZCTMTWRHEBIN-ANBDAQEENA-N
SMILES: OC(=O)[C@@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)