accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFMgBr

CAS RN 112981-69-8

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl](bromo)magnesium
Synonyms: [3,5-bis(trifluoromethyl)phenyl](bromo)magnesium
Molecular Weight (g/mol): 317.31
Molecular Formula: C8H3BrF6Mg
InChi Key: FTKNONULORTDRS-UHFFFAOYSA-M
SMILES: FC(F)(F)C1=CC(=CC([Mg]Br)=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results