Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Bis(trifluoromethyl)phenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC392151000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392151000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC392151000 Supplier Thermo Scientific Chemicals Supplier No. 392151000
  • $309.00
    $247.65 / Each of 1

    Save $61.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 112981-69-8 | C8H3BrF6Mg | 317.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112981-69-8 , 109-99-9
Molecular Formula C8H3BrF6Mg
Molecular Weight (g/mol) 317.31
MDL Number MFCD07369723
InChI Key FTKNONULORTDRS-UHFFFAOYSA-M
Synonym magnesium, 3,5-bis trifluoromethyl phenyl bromo, 3,5-bis trifluoromethyl phenylmagnesiumbromide, 3,5-bis trifluoromethyl phenylmagnesium bromide, 3,5-bis trifluoromethyl phenylmagnesium bromide solution, acmc-20mhbp, ftknonulortdrs-uhfffaoysa-m, 3,5-di trifluoromethyl phenylmagnesium bromide, grignard reagent
PubChem CID 11197896
IUPAC Name magnesium;1,3-bis(trifluoromethyl)benzene-5-ide;bromide
SMILES FC(F)(F)C1=CC(=CC([Mg]Br)=C1)C(F)(F)F

Specifications

CAS Min % 12.51, 81.24
CAS Max % 18.76, 87.5
Color Brown
Density 1.0150g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass bottle
Linear Formula BrMgC6H3(CF3)2
Quantity 100 mL
Specific Gravity 1.015
Formula Weight 317.31
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)phenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
May form explosive peroxides.
May cause respiratory irritation.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.