accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1131-16-4

IUPAC Name: 3,5-dimethyl-1-phenyl-1H-pyrazole
Synonyms: 3,5-dimethyl-1-phenylpyrazole
Molecular Weight (g/mol): 172.23
Molecular Formula: C11H12N2
InChi Key: ULPMPUPEFBDQQA-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)