accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNNNNFHClHCl

CAS RN 1134322-93-2

IUPAC Name: 1-{1-[(4-fluorophenyl)methyl]-1H-1,3-benzodiazol-2-yl}-N-methylpiperidin-4-amine dihydrochloride
Synonyms: 1-{1-[(4-fluorophenyl)methyl]-1,3-benzodiazol-2-yl}-N-methylpiperidin-4-amine dihydrochloride
Molecular Weight (g/mol): 411.35
Molecular Formula: C20H25Cl2FN4
InChi Key: ZIGFBTWAPHHQRA-UHFFFAOYSA-N
SMILES: Cl.Cl.CNC1CCN(CC1)C1=NC2=CC=CC=C2N1CC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results