CAS RN 113558-89-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ E-4031
A potent and selective inhibitor of the rapid type of inactivating inward-rectifying K+ current (IK, IR) (IC50 = 10 nM).