CAS RN 113559-13-0
IUPAC Name:
dihydrogen N-(4-{1-[2-(6-methylpyridin-2-yl)ethyl]piperidine-4-carbonyl}phenyl)methanesulfonamide dichloride
Synonyms:
dihydrogen N-(4-{1-[2-(6-methylpyridin-2-yl)ethyl]piperidine-4-carbonyl}phenyl)methanesulfonamide dichloride
Molecular Weight (g/mol):
474.44
Molecular Formula:
C21H29Cl2N3O3S
InChi Key:
ZQBNWMFBOSOOLX-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CC1=CC=CC(CCN2CCC(CC2)C(=O)C2=CC=C(NS(C)(=O)=O)C=C2)=N1
1
–
1
of
1
results
1
R&D Systems™ E-4031 dihydrochloride
CAS: 113559-13-0 Molecular Formula: C21H29Cl2N3O3S