CAS RN 113760-29-5
IUPAC Name:
6,6-dimethyl-2-(phenylimino)-4,5,6,7-tetrahydro-2H-1,3-benzoxathiol-4-one
Synonyms:
6,6-dimethyl-2-(phenylimino)-5,7-dihydro-1,3-benzoxathiol-4-one
Molecular Weight (g/mol):
273.35
Molecular Formula:
C15H15NO2S
InChi Key:
XFSSJIQCIPSBHJ-UHFFFAOYSA-N
SMILES:
CC1(C)CC2=C(SC(O2)=NC2=CC=CC=C2)C(=O)C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals NF-kB Activation Inhibitor VI, BOT-64
CAS: 113760-29-5 Molecular Formula: C15H15NO2S Synonym: (Z)-6,6-Dimethyl-2-(phenylimino)-6,7-dihydrobenzo[d][1,3]oxathiol-4(5H)-one