accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2POHHOOCl

CAS RN 114012-12-3

IUPAC Name: [3-amino-2-(4-chlorophenyl)propyl]phosphonic acid
Synonyms: phaclofen
Molecular Weight (g/mol): 249.63
Molecular Formula: C9H13ClNO3P
InChi Key: VSGNGLJPOGUDON-UHFFFAOYNA-N
SMILES: NCC(CP(O)(O)=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results