accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OHNOCH3OCH3OH3C

CAS RN 114289-47-3

IUPAC Name: 2,2-dimethyl-N-(2,4,6-trimethoxyphenyl)dodecanamide
Synonyms: 2,2-dimethyl-N-(2,4,6-trimethoxyphenyl)dodecanamide
Molecular Weight (g/mol): 393.57
Molecular Formula: C23H39NO4
InChi Key: WAFNZAURAWBNDZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC(C)(C)C(=O)NC1=C(OC)C=C(OC)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)