CAS RN 1143-70-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ Urolithin A
A Tocris product, part of R&D Systems. PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC)