accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3COSSOHOClN(E)

CAS RN 115104-28-4

IUPAC Name: 3-[({3-[(1E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl}({[2-(dimethylcarbamoyl)ethyl]sulfanyl})methyl)sulfanyl]propanoic acid
Synonyms: (R-(E))-verlukast (r-(e))-isomer verlukast (r-(e))-verlukast verlukast, (R-(E))- (R-(E))-isomer verlukast verlukast, (r-(e))-isomer verlukast, (R-(E))-isomer verlukast, (r-(e))-
Molecular Weight (g/mol): 515.08
Molecular Formula: C26H27ClN2O3S2
InChi Key: AXUZQJFHDNNPFG-UXBLZVDNNA-N
SMILES: CN(C)C(=O)CCSC(SCCC(O)=O)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results