CAS RN 1152197-23-3
IUPAC Name:
[(6-{6-amino-8-[(4-chlorophenyl)sulfanyl]-9H-purin-9-yl}-7-methoxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-yl)oxy]methyl acetate
Synonyms:
[(6-{6-amino-8-[(4-chlorophenyl)sulfanyl]purin-9-yl}-7-methoxy-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-yl)oxy]methyl acetate
Molecular Weight (g/mol):
557.9
Molecular Formula:
C20H21ClN5O8PS
InChi Key:
FZMWUFYPEVDWPA-UHFFFAOYNA-N
SMILES:
COC1C(OC2COP(=O)(OCOC(C)=O)OC12)N1C(SC2=CC=C(Cl)C=C2)=NC2=C(N)N=CN=C12
1
–
1
of
1
results
1
R&D Systems™ 8-pCPT-2-O-Me-cAMP-AM
CAS: 1152197-23-3 Molecular Formula: C20H21ClN5O8PS