accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFFFOHNOHO

CAS RN 1152782-19-8

IUPAC Name: 2-({4-[2-(trifluoromethyl)phenyl]piperidine-1-carbonyl}amino)benzoic acid
Synonyms: 2-{4-[2-(trifluoromethyl)phenyl]piperidine-1-carbonylamino}benzoic acid
Molecular Weight (g/mol): 392.38
Molecular Formula: C20H19F3N2O3
InChi Key: MEAQCLPMSVEOQF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1NC(=O)N1CCC(CC1)C1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)