accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOSOOO(S)O

CAS RN 115314-14-2

IUPAC Name: [(2S)-oxiran-2-yl]methyl 3-nitrobenzene-1-sulfonate
Synonyms: (2S)-oxiran-2-ylmethyl 3-nitrobenzenesulfonate
Molecular Weight (g/mol): 259.23
Molecular Formula: C9H9NO6S
InChi Key: AIHIHVZYAAMDPM-QMMMGPOBSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)