CAS RN 115338-32-4
IUPAC Name:
hydrogen 2-{4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl}-2,3-dihydro-1H-isoindole-1,3-dione bromide
Synonyms:
hydrogen 2-{4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl}isoindole-1,3-dione bromide
Molecular Weight (g/mol):
474.4
Molecular Formula:
C23H28BrN3O3
InChi Key:
AXRUEPFPTQYHQD-UHFFFAOYSA-N
SMILES:
[H+].[Br-].COC1=CC=CC=C1N1CCN(CCCCN2C(=O)C3=CC=CC=C3C2=O)CC1
1
–
1
of
1
results
1
R&D Systems™ NAN-190 hydrobromide
CAS: 115338-32-4 Molecular Formula: C23H28BrN3O3