accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH3NNH2NClONNH2H2N

CAS RN 1154-25-2

IUPAC Name: 3-amino-6-chloro-N-(diaminomethylidene)-5-[ethyl(propan-2-yl)amino]pyrazine-2-carboxamide
Synonyms: 3-amino-6-chloro-N-(diaminomethylidene)-5-[ethyl(isopropyl)amino]pyrazine-2-carboxamide
Molecular Weight (g/mol): 299.76
Molecular Formula: C11H18ClN7O
InChi Key: QDERNBXNXJCIQK-UHFFFAOYSA-N
SMILES: CCN(C(C)C)C1=NC(N)=C(N=C1Cl)C(=O)N=C(N)N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results