accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116-17-6

IUPAC Name: tris(propan-2-yl) phosphite
Synonyms: triisopropyl phosphite
Molecular Weight (g/mol): 208.24
Molecular Formula: C9H21O3P
InChi Key: SJHCUXCOGGKFAI-UHFFFAOYSA-N
SMILES: CC(C)OP(OC(C)C)OC(C)C
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (4)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)