Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropyl Phosphite (Tech.), 90%, Thermo Scientific™

Catalog No. AC139875000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC139875000 500 g
AC139871000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC139875000 Supplier Thermo Scientific Chemicals Supplier No. 139875000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-17-6
Molecular Formula C9H21O3P
Molecular Weight (g/mol) 208.238
InChI Key SJHCUXCOGGKFAI-UHFFFAOYSA-N
Synonym triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite
PubChem CID 8304
IUPAC Name tripropan-2-yl phosphite
SMILES CC(C)OP(OC(C)C)OC(C)C

Specifications

Color Colorless
Density 0.914
Infrared Spectrum Authentic
Refractive Index 1.4094 to 1.4114 (20°C, 589nm)
Linear Formula [(CH3)2CHO]3P
Quantity 500 g
Beilstein 01, 363
Fieser 01,1229; 16,360
Formula Weight 208.24
Percent Purity ≥88%
Grade Technical
Physical Form Liquid
Chemical Name or Material Triisopropyl phosphite, tech., 90%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Hazard Statement AIR SENSITIVE
EINECSNumber 204-130-0
RTECSNumber TH2800000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.