Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropyl Phosphite, TCI America™
SDP

Catalog No. T061025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T061025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T061025ML Supplier TCI America Supplier No. T061025ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 116-17-6
Molecular Formula C9H21O3P
Molecular Weight (g/mol) 208.238
MDL Number MFCD00008874
InChI Key SJHCUXCOGGKFAI-UHFFFAOYSA-N
Synonym triisopropyl phosphite, tri-2-propyl phosphite, phosphorous acid, tris 1-methylethyl ester, isopropyl phosphite, tri, triisopropoxyphosphine, phosphorous acid, triisopropyl ester, tri-2-propylphosphite, unii-rj7qu4yqun, triisopropylphosphite, tri-i-propylphosphite
PubChem CID 8304
IUPAC Name tripropan-2-yl phosphite
SMILES CC(C)OP(OC(C)C)OC(C)C

Specifications

Color Colorless
Boiling Point 74°C
UN Number 3278
Quantity 25 mL
Formula Weight 208.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triisopropyl Phosphite
RTECSNumber TH2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.