CAS RN 116-63-2
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.25 MDL Number: MFCD00004019 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N Synonym: 1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine PubChem CID: 8316 ChEBI: CHEBI:19024 IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.245 MDL Number: MFCD00004019 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N Synonym: 1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine PubChem CID: 8316 ChEBI: CHEBI:19024 IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 99%
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.245 MDL Number: MFCD00004019 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N Synonym: 1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine PubChem CID: 8316 ChEBI: CHEBI:19024 IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
4-Amino-3-hydroxy-1-naphthalene Sulfonic Acid, ACS, 90%, Spectrum™ Chemical
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.25 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: NC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.25 MDL Number: MFCD00004019 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N Synonym: 1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine PubChem CID: 8316 ChEBI: CHEBI:19024 IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
1-Amino-2-napthol-4-sulfonic Acid, ACS Reagent Grade, Ricca Chemical
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.245 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N Synonym: 1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine PubChem CID: 8316 ChEBI: CHEBI:19024 IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
1-Amino-2-Naphthol-4-Sulfonic Acid, Reagent Grade, LabChem™
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.245 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N Synonym: 1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine PubChem CID: 8316 ChEBI: CHEBI:19024 IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
1-Amino-2-naphthol-4-sulfonic Acid 97.0+%, TCI America™
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.245 MDL Number: MFCD00004019 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N Synonym: 1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine PubChem CID: 8316 ChEBI: CHEBI:19024 IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
Avantor Macron 1-Amino-2-Naphthol-4-Sulfonic Acid, OR, Macron Fine Chemicals™
CAS: 116-63-2 Molecular Formula: C10H9NO4S Molecular Weight (g/mol): 239.245 InChI Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N Synonym: 1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine PubChem CID: 8316 ChEBI: CHEBI:19024 IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid SMILES: C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O