Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-hydroxy-1-naphthalene Sulfonic Acid, ACS, 90%, Spectrum™ Chemical
SDP

Catalog No. 18608261 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-261 5 kg Poly Pail
18-608-257 5 g Amber Glass Bottle
18-608-258 25 g Poly Bottle
18-608-259 100 g Poly Bottle
18-608-260 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-261 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A11165KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

CAS: 116-63-2 | Formula: C10H9NO4S | Weight: 239.25 g/mol

Chemical Identifiers

CAS 116-63-2
Molecular Formula C10H9NO4S
Molecular Weight (g/mol) 239.25
InChI Key RXCMFQDTWCCLBL-UHFFFAOYSA-N
IUPAC Name 4-amino-3-hydroxynaphthalene-1-sulfonic acid
SMILES NC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Poly Pail
Quantity 5 kg
Percent Purity 0.9
Grade ACS

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.