accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNNH2NClOHNNH2HN

CAS RN 1161-94-0

IUPAC Name: {[(3,5-diamino-6-chloropyrazin-2-yl)formamido](phenylamino)methylidene}azanium
Synonyms: {[(3,5-diamino-6-chloropyrazin-2-yl)formamido](phenylamino)methylidene}azanium
Molecular Weight (g/mol): 306.73
Molecular Formula: C12H13ClN7O
InChi Key: NIOHELZQFBGCEO-UHFFFAOYSA-O
SMILES: NC1=NC(N)=C(N=C1Cl)C(=O)NC(=[NH2+])NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)