accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 116853-97-5

IUPAC Name: 2-chloro-6-methoxypyridine-4-carbonyl chloride
Synonyms: 2-chloro-6-methoxypyridine-4-carbonyl chloride
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: RITCMXVSFUDYGY-UHFFFAOYSA-N
SMILES: COC1=NC(Cl)=CC(=C1)C(Cl)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results