accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117-83-9

IUPAC Name: 1,2-bis(2-butoxyethyl) benzene-1,2-dicarboxylate
Synonyms: palatinol dibutylglycol phthalate palatinol K di(butoxyethyl)phthalate phthalic acid dibutoxy ethyl ester kronisol kesscoflex butyl "cellosolve" phthalate dibutyl cellosolve phthalate bis(2-butoxyethyl)phthalate
Molecular Weight (g/mol): 366.45
Molecular Formula: C20H30O6
InChi Key: CMCJNODIWQEOAI-UHFFFAOYSA-N
SMILES: CCCCOCCOC(=O)C1=CC=CC=C1C(=O)OCCOCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results