Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalic Acid Bis(2-butoxyethyl) Ester 85.0+%, TCI America™
SDP

Catalog No. P065725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P065725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P065725G Supplier TCI America Supplier No. P065725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117-83-9
Molecular Formula C20H30O6
Molecular Weight (g/mol) 366.454
MDL Number MFCD00048781
InChI Key CMCJNODIWQEOAI-UHFFFAOYSA-N
Synonym Bis(2-butoxyethyl) Phthalate
PubChem CID 8345
ChEBI CHEBI:79937
IUPAC Name bis(2-butoxyethyl) benzene-1,2-dicarboxylate
SMILES CCCCOCCOC(=O)C1=CC=CC=C1C(=O)OCCOCCCC

Specifications

Color Yellow
Boiling Point 270°C
Quantity 25 g
Formula Weight 366.45
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Phthalic Acid Bis(2-butoxyethyl) Ester
EINECSNumber (3)-2962
RTECSNumber TI0175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.