accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OCH3OH3CHO(E)

CAS RN 117048-59-6

IUPAC Name: 2-methoxy-5-[(1E)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenol
Synonyms: combretastatin A-4
Molecular Weight (g/mol): 316.35
Molecular Formula: C18H20O5
InChi Key: HVXBOLULGPECHP-AATRIKPKSA-N
SMILES: COC1=CC=C(\C=C\C2=CC(OC)=C(OC)C(OC)=C2)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results