Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Combretastatin A4 98.0+%, TCI America™
SDP

Catalog No. C2520250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2520250MG 250 mg
C252025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2520250MG Supplier TCI America Supplier No. C2520250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 117048-59-6
Molecular Formula C18H20O5
Molecular Weight (g/mol) 316.353
MDL Number MFCD03453309
InChI Key HVXBOLULGPECHP-WAYWQWQTSA-N
PubChem CID 5351344
IUPAC Name 2-methoxy-5-[(Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenol
SMILES COC1=C(C=C(C=C1)C=CC2=CC(=C(C(=C2)OC)OC)OC)O

Specifications

Melting Point 83°C
Color White
Quantity 250 mg
Synonym (Z)-2-Methoxy-5-(3,4,5-trimethoxystyryl)phenol, 3′-Hydroxy-3,4,4′,5-tetramethoxy-cis-stilbene
Formula Weight 316.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Combretastatin A4
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.