accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHFH3C(E/Z)(E/Z)

CAS RN 1170613-55-4

IUPAC Name: N-[1-(4-fluorophenyl)-2-methylpent-1-en-3-ylidene]hydroxylamine
Synonyms: N-[1-(4-fluorophenyl)-2-methylpent-1-en-3-ylidene]hydroxylamine
Molecular Weight (g/mol): 207.25
Molecular Formula: C12H14FNO
InChi Key: HKROEBDHHKMNBZ-UHFFFAOYSA-N
SMILES: CCC(=NO)C(C)=CC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)