CAS RN 1173022-16-6
IUPAC Name:
1'-{2-[4-(trifluoromethyl)phenyl]ethyl}-1,2-dihydrospiro[3,1-benzoxazine-4,4'-piperidin]-2-one hydrochloride
Synonyms:
1'-{2-[4-(trifluoromethyl)phenyl]ethyl}-1H-spiro[3,1-benzoxazine-4,4'-piperidin]-2-one hydrochloride
Molecular Weight (g/mol):
426.86
Molecular Formula:
C21H22ClF3N2O2
InChi Key:
KRRISOFSWVKYBF-UHFFFAOYSA-N
SMILES:
Cl.FC(F)(F)C1=CC=C(CCN2CCC3(CC2)OC(=O)NC2=CC=CC=C32)C=C1
1
–
1
of
1
results
1
R&D Systems™ RS 102895 hydrochloride
CAS: 1173022-16-6 Molecular Formula: C21H22ClF3N2O2